Learn More
BRAND™ Class B Polypropylene Volumetric Flask
Product Code. 17470134 Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 2
Product Code. 17470134
Brand: BRAND™ 36054
High clarity polypropylene construction. Exposure to temperatures up to 60°C will not cause permanently exceeded error limits.
  • High clarity polypropylene construction
  • Includes polypropylene stopper
  • Error limits according to class B, DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • Exposure to temperatures up to 60°C (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60°C is recommended to preserve marks and inscriptions
  • Height = Height without stopper
TRUSTED_SUSTAINABILITY
Specifications
Calibration To contain (TC, in.)
Certifications/Compliance DIN EN ISO 1042
Closure Material Polypropylene
Capacity (Metric) 500 mL
Material Polypropylene
Height (Metric) 258 mm
Limit of Error ±0.5 mL
No. per Pack 1
Temperature Range (Metric) Up to 60°C
Tolerance ±0.5 mL
Autoclavable Autoclavable
Class B
Closure Type Stopper
Closure Size NS 19/26