Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes
Product Code. p-8013421
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
4 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11732252
Brand: Hirschmann™ 1300129
Only null left
Add to basket
Add to basket
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
  • One mark
  • Amber stain graduation
  • Acc. to DIN EN ISO 648
Specifications
Class B
Material Glass
Graduated Yes
Graduation Color Brown
Quantity 12/Pk.
Tolerance 0.08 mL
Volume (Metric) 0.5 mL
Length (Metric) 300 mm
Type Volumetric Pipet