Learn More
VITLAB™ Class A PMP Graduated Cylinders with Raised Scale
Click to view available options
Graduations:
0.5 mL
1 mL
Unit Size:
Pack of 2
Specifications
Specifications
| Class | A |
| Graduations | 1 mL |
| Limit of Error | ±0.5mL |
| Shape | Round |
| For Use With (Application) | Food Industry and Volume Measurement |
| Base Type | Hexagonal |
| Height (Metric) | 200 mm |
| Type | Graduated Cylinder |