Learn More
OHAUS™ Clamp Double
Shop All Ohaus Products
Click to view available options
Dimensions (L x W x H):
30.000 x 20.000 x 5.000 cm
Unit Size:
1 unit
Description
- Overhead Stirrer Accessory
Specifications
Specifications
| Height (Metric) | 5.000 cm |
| Width (Metric) | 20.000 cm |
| Weight (Metric) | 200.000 g |
| Dimensions (L x W x H) | 30.000 x 20.000 x 5.000 cm |