missing translation for 'onlineSavingsMsg'
Learn More

Cladribine, Mikromol™

Product Code. 17989699
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989699 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17989699 Supplier Mikromol Supplier No. MM3381.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cladribine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cladribine
CAS 4291-63-8
Analyte or Component Names Cladribine
Shipping Condition Room Temperature
Molecular Formula C10 H12 Cl N5 O3
Certifications/Compliance ISO/IEC 17025
Synonym Cladribine, Adenosine, 2-chloro-2'-deoxy-, 2-Chloro-2'-deoxyadenosine, (2R,3S,5R)-5-(6-Amino-2-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol, 2-CdA, 2-Chlorodeoxyadenosine, Biodribin, Cladarabine, CldAdo, Jk 6251, Leustat, Leustatin, NSC 105014, NSC 105014-F, RWJ 26251, 2-Chloro-9-(2-deoxy-beta-d-erythro-pentofuranosyl)-9H-purin-6-amine, 2-Chloro-2'-deoxy-beta-adenosine, 2-Chloro-6-amino-9-(2-deoxy-beta-D-erythro-pentofuranosyl)purine
InChI Formula InChI=1S/C10H12ClN5O3/c11-10-14-8(12)7-9(15-10)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H2,12,14,15)/t4-,5+,6+/m0/s1
SMILES Nc1nc(Cl)nc2c1ncn2[C@H]3C[C@H](O)[C@@H](CO)O3
IUPAC Name (2R,3S,5R)-5-(6-amino-2-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol) 285.69
Quantity 250 mg
Formula Weight 285.0629 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.