missing translation for 'onlineSavingsMsg'
Learn More

Citronellol, 95%

Product Code. p-7024410
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11429326

Brand: Thermo Scientific Alfa Aesar A19016.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-22-9 | C10H20O | 156.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-22-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002935
InChI Key QMVPMAAFGQKVCJ-UHFFFAOYSA-N
Synonym citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
PubChem CID 8842
ChEBI CHEBI:50462
IUPAC Name 3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO

Specifications

Melting Point <-20°C
Density 0.858
Boiling Point 221°C to 222°C
Flash Point 98°C (208°F)
Odor Pleasant
Refractive Index 1.4575
Quantity 100 g
UN Number UN3082
Beilstein 1721507
Merck Index 14,2330
Formula Weight 156.27
Percent Purity 95%
Chemical Name or Material Citronellol
Show More Show Less
compliance-icons
Product Identifier
  • Citronellol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Citronellol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.