missing translation for 'onlineSavingsMsg'
Learn More

Citicoline-d9 Sodium Salt, TRC

Product Code. 30360665
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360665 0.5 mg 0.5mg
30349253 10 mg 10mg
30355402 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360665 Supplier Toronto Research Chemicals Supplier No. TRCC5080520.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Citicoline-d9 Sodium Salt
Molecular Formula C14 D9 H16 N4 Na O11 P2
Quantity 0.5 mg
Synonym Cytidine 5'-(Trihydrogen Diphosphate) P'-[2-(Trimethylammonio-d9)ethyl] Ester Inner Salt Sodium Salt, Acticolin-d9, Brassel-d9, Cebroton-d9, Cidifos-d9, Citicoline-d9 Sodium, Flussorex-d9, Gerolin-d9, IP 302-d9 Sodium, Logan-d9, Neurotron-d9, Sinkron-d9
InChI Formula InChI=1S/C14H26N4O11P2.Na/c1-18(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)17-5-4-10(15)16-14(17)21;/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H3-,15,16,21,22,23,24,25);/q;+1/p-1/t9-,11-,12-,13-;/m1./s1/i1D3,2D3,3D3;
SMILES [2H]C([2H])([2H])[N+](CCOP(=O)(O[Na])OP(=O)([O-])OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=NC2=O)N)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name [[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-[2-[tris(trideuteriomethyl)azaniumyl]ethoxy]phosphoryl]oxysodium
Molecular Weight (g/mol) 519.361
Formula Weight 519.146
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.