missing translation for 'onlineSavingsMsg'
Learn More

cis-4,7,10,13,16,19-Docosahexaenoic Acid, >99%, MP Biomedicals™

Product Code. 11451331 Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
25 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
25mg
This item is not returnable. View return policy

Product Code. 11451331

missing translation for 'mfr': MP Biomedicals 0215909710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00065722
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Color Colorless
Assay Percent Range ≥98%
Quantity 10 mg
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
Formula Weight 328.5
Physical Form Liquid
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.