missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15457246
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457246 250 mg 250mg
15438828 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457246 Supplier Thermo Scientific Alfa Aesar Supplier No. H62281.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 222530-33-8
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD01862295
InChI Key JSGHMGKJNZTKGF-BDAKNGLRSA-N
Synonym 1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
PubChem CID 16218560
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 250 mg
Formula Weight 243.29
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.