Learn More
Ciliobrevin A, MedChemExpress
Product Code. 16479716
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16479716
Brand: Medchem Express HY100790
MedChemExpress Ciliobrevin A (HPI-4) is a hedgehog (Hh) signaling pathway inhibitor with median inhibitory concentration (IC50) less than 10 μM.
Ciliobrevin A (HPI-4) is a hedgehog (Hh) signaling pathway inhibitor with median inhibitory concentration (IC50) less than 10 μM.
Specifications
Chemical Name or Material Ciliobrevin A
Physical Form Solid
CAS 302803-72-1
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H9Cl2N3O2
For Use With (Application) Cancer-Kinase/protease
Synonym HPI-4
Solubility Information DMSO : 100 mg/mL (279.19 mM; Need ultrasonic)
SMILES N#C/C(C(C1=CC=C(Cl)C=C1Cl)=O)=C(N2)\NC3=C(C=CC=C3)C2=O
Molecular Weight (g/mol) 358.18
Formula Weight 358.18
Percent Purity 98.57%
Grade Research