missing translation for 'onlineSavingsMsg'
Learn More

CIL56, MedChemExpress

Product Code. 16497267
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497267 5 mg 5mg
16460857 50 mg 50mg
16455406 100 mg 100mg
16443126 25 mg 25mg
16434558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497267 Supplier Medchem Express Supplier No. HY112063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CIL56 is a potent and selective ferroptosis inducer. Ferroptosis is an iron-dependent form of regulated cell death (RCD).

CIL56 is a potent and selective ferroptosis inducer. Ferroptosis is an iron-dependent form of regulated cell death (RCD).

Specifications

Chemical Name or Material CIL56
CAS 300802-28-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27N3O5S2
Quantity 5 mg
Solubility Information DMSO : 41.67 mg/mL (85.11 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC2=C(C=C1)C3=C(C=C(S(=O)(N4CCCCC4)=O)C=C3)/C2=N\O)(N5CCCCC5)=O
Molecular Weight (g/mol) 489.61
Formula Weight 489.61
Percent Purity 97.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.