missing translation for 'onlineSavingsMsg'
Learn More

CID755673, MedChemExpress

Product Code. 16431277
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431277

Brand: Medchem Express HY12239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CID755673 is a potent PKD inhibitor with IC50s of 182 nM, 280 nM and 227 nM for PKD1, PKD2 and PKD3, respectively.

CID755673 is a potent PKD inhibitor with IC50s of 182 nM, 280 nM and 227 nM for PKD1, PKD2 and PKD3, respectively.

Specifications

Chemical Name or Material CID755673
CAS 521937-07-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H11NO3
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (460.36 mM; Need ultrasonic)
SMILES O=C1NCCCC2=C1OC3=CC=C(O)C=C32
Molecular Weight (g/mol) 217.22
Formula Weight 217.22
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.