missing translation for 'onlineSavingsMsg'
Learn More

CID 1375606, MedChemExpress

Product Code. p-200100433
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464788 10 mg 10mg
16441057 50 mg 50mg
16417537 5 mg 5mg
16413246 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464788 Supplier Medchem Express Supplier No. HY114146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27.

CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27.

Specifications

Chemical Name or Material CID 1375606
Physical Form Solid
CAS 313493-80-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H14Cl2N2O2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 53.33 mg/mL (138.43 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(NC2=CC=CC=C2)=O)C=C1)C3=CC=C(Cl)C=C3Cl
Molecular Weight (g/mol) 385.24
Formula Weight 385.24
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.