Learn More
CID 1375606, MedChemExpress
Product Code. 16464788
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464788 10 mg 10mg
16441057 50 mg 50mg
16417537 5 mg 5mg
16413246 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16464788 Supplier Medchem Express Supplier No. HY114146
MedChemExpress CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27.
CID 1375606 is a surrogate agonist of orphan G protein-coupled receptor GPR27.
Specifications
Chemical Name or Material CID 1375606
Physical Form Solid
CAS 313493-80-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H14Cl2N2O2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 53.33 mg/mL (138.43 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(NC2=CC=CC=C2)=O)C=C1)C3=CC=C(Cl)C=C3Cl
Molecular Weight (g/mol) 385.24
Formula Weight 385.24
Percent Purity 98.0%
Grade Research