missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHST2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16637964
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16637964 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637964 Supplier Invitrogen™ Supplier No. PA5101486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHST2.

N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHST2
Gene Accession No. Q9Y4C5
Gene Alias AI428561; AW121776; C130041E03Rik; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2; carbohydrate sulfotransferase 2; Chst2; Chts2; epididymis secretory protein Li 75; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; GlcNAc6ST; glcNAc6ST-1; GN6ST; Gn6ST-1; GST2; GST-2; HEL-S-75; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine-6-O-sulfotransferase
Gene Symbols CHST2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHST2(Accession Q9Y4C5), corresponding to amino acid residues P10-A60.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9435
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.