missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHST10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16673065 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16673065 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16673065 Supplier Invitrogen™ Supplier No. PA5106630

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHST10.

This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.[provided by RefSeq, Feb 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHST10
Gene Accession No. O43529, O54702, Q6PGK7
Gene Alias AI507003; AU041319; carbohydrate sulfotransferase 10; CHST10; HNK-1 sulfotransferase; HNK1ST; HNK-1ST; huHNK-1ST; raHNK-1ST; ST; sul-T
Gene Symbols CHST10
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHST10(Accession O43529), corresponding to amino acid residues K200-R250.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140568, 9486, 98388
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.