Learn More
RETSCH Chrome Steel Comfort Grinding Jars
Product Code. p-8003616 Shop All Retsch Products
Click to view available options
Capacity (Metric):
50 mL
125 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 10402272
Brand: RETSCH 01.462.0224
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these grinding jars with Retsch™ ball mills. Retsch™ Chrome Steel Comfort Grinding Jars feature an O-type sealing for safe operation.
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Product Type Comfort Grinding Jar
Capacity (Metric) 250 mL
For Use With (Equipment) For PM 100, PM 400 and S 100
Material Chrome Steel