Specifications
| Base Material | Weakly Acidic Silica |
| Description | Adsorbent Weight: 420 mg |
| For Use With (Application) | Aflatoxins, Chloramphenicol, Pesticides, Steroids, Vitamins |
| Particle Size | 45 μm |
| Sorbent | SiOH |
| Type | Chromafix SiOH Cartridge |
| Pore Size | 60 Å |
| Quantity | 50 Pack |
| Surface Area (Metric) | 500 m2/g |