Learn More
Macherey-Nagel™ Chromabond™ Glass Fiber Frit
Shop All Macherey-Nagel Products
Click to view available options
Volume (Metric):
3 mL
6 mL
Unit Size:
Pack of 250
Description
Frits act as filters in solid phase extraction chromatography. They help to hold the column packing in place and facilitates sorbent packing in the column. Glass fiber columns/frits have many advantages:
- Resistant to acids, bases and most organic solvents
- Has low absorption for most biological macromolecules
- Ideal for high-temperature applications
Specifications
Specifications
| For Use With (Application) | Solid Phase Extraction |
| For Use With (Equipment) | Glass Column |
| Type | Frit |
| Volume (Metric) | 3 mL |
| Material | Glass Fiber |
| Quantity | 250 Pack |