Description
- Automated systems for CHROMABOND™ vacuum manifolds
- Polypropylene column, polyethylene filter element
- High loading capacity and separation efficiency
- Constant back pressure, airtight sealing
- Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
Specifications
| Base Material | Silica |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 1000 mg |
| For Use With (Application) | Solid Phase Extraction |
| Sorbent | Chromabond C18 ec |
| Type | Solid Phase Extraction Column |