Learn More
Macherey-Nagel™ Chromabond™ CN/SiOH Glass SPE Column
Product Code. 12817991 Shop All Macherey-Nagel Products
Change view
Click to view available options
Column Capacity (Metric):
6 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity (Metric) unitSize
12817991 6 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12817991 Supplier Macherey-Nagel™ Supplier No. 730135G
Specifications
Column Capacity (Metric) 6 mL
Column Type SPE
For Use With (Application) Solid Phase Extraction Analysis of PAH
Sorbent Chromabond CN/SiOH
Type Solid Phase Extraction Column
Functionality/Interaction Type p-p Interaction