Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| For Use With (Application) | Solid Phase Extraction Analysis of PAH |
| Sorbent | Chromabond CN/SiOH |
| Type | Solid Phase Extraction Column |
| Functionality/Interaction Type | p-p Interaction |