Learn More
Macherey-Nagel™ Chromabond™ C18 ec Gerstel™ MPS Column
Product Code. 12857851 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Column Capacity (Metric) unitSize
12857851 500 mg 6 mL Each
12804391 500 mg 3 mL Each
12881061 200 mg 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12857851 Supplier Macherey-Nagel™ Supplier No. 730013MPS
Specifications
Column Capacity (Metric) 6 mL
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Solid Phase Extraction
Type Solid Phase Extraction Column