Learn More
Azpack™ Freeze Drying and Injection Vials
Product Code. 10653205
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
Diameter (Metric):
14.25 mm (Neck O.D.)
22.25 mm (Neck O.D.)
24.25 mm (Neck O.D.)
29.25 mm (Neck O.D.)
Height (Metric):
38 mm
39.5 mm
46 mm
56 mm
Unit Size:
Pack of 104
Pack of 154
Pack of 186
Pack of 441
Product Code. 10653205
Brand: Azpack™ 10653205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure optimum protection of the contents against premature aging. Azpack™ Freeze Drying and Injection Vials protect content integrity and loss of effectiveness.
  • Manufactured from FIOLAX™ neutral clear glass
  • Outstanding chemical resistance, neutrality and impermeability
  • Crimp neck
  • Supplied without caps
Specifications
Type Vial
Capacity (Metric) 2 mL
Color Clear
Material FIOLAX™ Glass
Neck Style Crimp
Quantity 441/Pk.
Closure Size 13mm
Diameter (Metric) 14.25 mm (Neck O.D.)
Diameter (Metric) Inside Neck 6.35 mm
Height (Metric) 39.5 mm