missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chondrolectin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109534
This item is not returnable.
View return policy
Description
Immunogen sequence: YRNWYTDEPS CGSEKCVVMY HQPTANPGLG GPYLYQWNDD RCNMKHNYIC KYEPEINPTA PVEKPYLTNQ PGDTHQNVVV TEAGIIPNLI YVVIPTIPLL L.
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
| Chondrolectin | |
| Polyclonal | |
| Unconjugated | |
| CHODL | |
| 3110074E07Rik; C21orf68; CHODL; Chondrolectin; FLJ12627; LOW QUALITY PROTEIN: chondrolectin; MT75; PRED12; Transmembrane protein MT75; UNQ872/PRO1890 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246048, 288289 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9CXM0 | |
| CHODL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-210 of human CHODL (NP_001191103.1). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction