missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine-d6 Maleate Salt, TRC

Product Code. 30041344
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041344 2.5 mg 2.5mg
30040997 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30041344 Supplier Toronto Research Chemicals Supplier No. TRCC4243032.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219806-45-7
Molecular Formula C16 D6 H13 Cl N2 . C4 H4 O4
Molecular Weight (g/mol) 396.8976
Synonym Chlorphenamine-D6 Maleate, Chlorpheniramine D6 Maleate, 3-(4-Chlorophenyl)-3-pyridin-2-yl-N,N-bis(trideuteriomethyl)propan-1-amine maleate, Chlorpheniramine-d6 maleate, Chlorphenamine-d6 maleate, Chlorphenamine-D6 (N,N-dimethyl-D6) Maleate, γ-(4-Chlorophenyl)-N,N-di(methyl-d3)-2-pyridinepropanamine (2Z)-2-Butenedioate Salt (1:1), 3-(4-Chlorophenyl)-N,N-di(methyl-d3)-3-(pyridin-2-yl)propan-1-amine (2Z)-2-Butenedioate
IUPAC Name (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-3-pyridin-2-yl-N,N-bis(trideuteriomethyl)propan-1-amine
SMILES [2H]C([2H])([2H])N(CCC(c1ccc(Cl)cc1)c2ccccn2)C([2H])([2H])[2H].OC(=O)\C=C/C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i1D3,2D3;
Formula Weight 396.1723
Percent Purity >95%
Chemical Name or Material Chlorphenamine-D6 Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.