missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine-d4 Phosphate Salt, TRC

Product Code. 30364012
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364012 10 mg 10mg
30361030 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30364012 Supplier Toronto Research Chemicals Supplier No. TRCC37996710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroquine-d4 Phosphate Salt
Purity Grade Notes HPLC
Molecular Formula C18H22D4ClN3 •xH3PO4
Quantity 10 mg
Synonym N4-(7-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine-d4 Phosphate Salt, Aralen-d4 Phosphate Salt, Aralen-d4 Phosphate, Arechin-d4, Avloclor-d4, Bemaphate-d4, Chingamin-d4, Chingamin-d4 Phosphate, SN 7618-d4, Sanoquin-d4, Tanakan-d4
InChI Formula InChI=1S/C18H26ClN3/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21)/i6D2,12D2
SMILES ClC1=CC=C2C(N=CC=C2NC(CC([2H])(C([2H])(N(CC)CC)[2H])[2H])C)=C1
IUPAC Name 4-N-(7-chloroquinolin-4-yl)-1,1,2,2-tetradeuterio-1-N,1-N-diethylpentane-1,4-diamine
Molecular Weight (g/mol) 323.9098
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.