missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Ethanol Stabilized, Multisolvent™ Grade, Sharlab

Product Code. 11308374
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11308374 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11308374 Supplier Scharlab Supplier No. CL02182500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Can be used for almost every laboratory application. Chloroform, Ethanol Stabilized, Multisolvent™ Grade from Sharlab features high purity and low water content for use in applications requiring an HPLC-grade, ACS-grade, ISO-grade, UV-VIS-grade or Reagent-grade product.

Multisolvent™ products are most frequently use for HPLC, but can also be used for numerous other applications, making it one of the most universal grades available. These versatile products have numerous advantages over application-specific solvents:

  • Guarantee results: QC controls are performed by modern analytical techniques for all guaranteed applications
  • Optimize purchasing and stocking: One product for most applications prevents the need to order, stock and track the supply of multiple grade
  • Optimal packaging: Highly-resistant and durable amber glass bottles or stainless steel drums ensure the highest product safety and quality
  • Lower costs by unifying: Multiple-use products at single-grade prices to avoid buying small quantities of specific products
  • Preserve the environment: Returnable packaging (one-way glass bottles or SS returnable drums) help prevent waste

Specifications

Melting Point -63°C
Absorbance 0.301 AU at 250nm, 0.046 AU at 265nm, 0.009 AU at 300nm
CAS 67-66-3
Color Colorless
Acidity Max. 0.0001meq/g
Appearance Clear
Density 1.474 - 1.483g/cm3
Quantity 2.5 L
Identification Passes test
Ignition Point 982°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.0002%
Molecular Formula CHCl3
Vapor Pressure 213 hPa
MDL Number MFCD00000826
UN Number 1888
Water Max. 0.01%
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Boiling Point 61°C
Solubility Information 8g/L in water
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES ClC(Cl)Cl
IUPAC Name trichloromethane
Molecular Weight (g/mol) 119.37
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38g/mol
Percent Purity Min. 99.8%
Chemical Name or Material Chloroform
Grade Applications: solvents, analytical chemistry, in the rubber industry
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with ethonol, Multisolvent HPLC grade ACS IOS UV-VIS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.