Learn More
JT Baker 2,5LT CHLOROFORM BAKER ANALYZED Reagent
Brand:  JT Baker 9180.2500
2,5LT CHLOROFORM BAKER ANALYZED Reagent
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 13295493
  • ONLINE EXCLUSIVE PRICE
    350.00 EUR
    167.56 EUR / 1 L
    Save 182.44 EUR
    52% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P281-Use personal protective equipment as required.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
SDS
Documents
Documents
Certificates
Promotions
Promotions