Learn More
WTW™ Chlorine Test Kits
Click to view available options
Type:
Cell Test Kit
Powder Pillows Kit
Reagent Test Kit
Range:
0.01-6.0 Cl2, Free/Total
0.02-2.0 mg/L Cl2, Free
0.02-2.0 mg/L Cl2, Total
0.03-6.0 Cl2 Free
0.03-6.0 Cl2, Free/Total
0.5-5.0 mg/L Cl2, Free
Unit Size:
Each
Pack of 100
Pack of 150
Specifications
Specifications
| Certifications/Compliance | DIN 38408 G4, EN ISO 7393 |
| Color Indicator | White |
| pH Range | 5 at 20°C |
| Volume (Metric) | 5 mL |
| For Use With (Application) | For disinfectants, waste water, drinking water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| Parameter Specifications | Chlorine |
| Range | 0.03-6.0 Cl2 Free |
| Storage Requirements | 15°C to 25°C |
| Show More |
Safety and Handling
Product Identifier
- chlorine cell test (free chlorine)
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : boric acid, ethylenedinitrilotetraaceticacid, tetrasodium salt