Learn More
Lovibond™ Chlorine test kit AF 112A
Product Code. 15944442
Click to view available options
Range:
0.1 to 1.0 mg/l Cl2
Unit Size:
Each
Product Code. 15944442
Brand: Lovibond™ 411120
Only null left
Add to basket
Add to basket
Chlorine test kit AF 112A featuring Comparator 2000+ measures 0.1 - 1.0 mg/l Cl2 includes case reagents and cells
The Comparator 2000+ is a versatile colorimetric system for water analysis. Comfortable handling, no compromise on accuracy and reproducibility.
Usable in different applications
The Comparator 2000+ is versatile: it is used in swimming pools as well as in research centers, universities or general drinking water treatment.
Daylight lamp
So that you can always measure safely and stably under unfavorable visibility conditions, we recommend the use of the (optional) mains or battery-powered daylight lamp. So the lighting conditions always remain the same. No matter where you are measuring.
Various analyses possible
For the Comparator 2000+, more than 400 color discs and numerous reagents are available. Your combination is guaranteed. And if you have to change them once, it works without opening the comparator.
Integrated prism
The integrated prism visually brings the glass standards of the color disc and the colored sample into view.
Specifications
Analytes Chlorine
Range 0.1 to 1.0 mg/l Cl2
Type Test Kit