Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Click to view available options
Type:
Chlorine Reagent
Liquid Reagent Kit
Unit Size:
Each
Description
- Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers
- High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
- Reagents follow an adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
- The methods use a DPD reagent in which the reaction between chlorine and reagent causes a pink tint in the sample
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
Specifications
Specifications
| No. of Tests | 300 |
| Quantity | 1 Each |
| Range | 0 to 5 mg/L |
| Test Method | adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G |
| Type | Chlorine Reagent |
Safety and Handling
Product Identifier
- Flüssig-Reagenz für 300 Tests Gesamtchlor
- Danger
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
- MIXTURE LIST-Contains : sulphuric acid, sodium hydroxide, potassium iodide