Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Product Code. 10204041
Click to view available options
Type:
Chlorine Reagent
Liquid Reagent Kit
Unit Size:
Each
Product Code. 10204041
Brand: Hanna Instruments™ HI93701T
Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers.
  • Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers
  • High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
  • Reagents follow an adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
  • The methods use a DPD reagent in which the reaction between chlorine and reagent causes a pink tint in the sample
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
Specifications
No. of Tests 300
Quantity 1 Each
Range 0 to 5 mg/L
Test Method adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
Type Chlorine Reagent
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Flüssig-Reagenz für 300 Tests Gesamtchlor
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, sodium hydroxide, potassium iodide