missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHL-1/L1CAM-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 EUR - 326.00 EUR
Specifications
| Antigen | CHL-1/L1CAM-2 |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642216
|
Novus Biologicals
NBP2-98946-100ul |
100 μg |
326.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623556
|
Novus Biologicals
NBP2-98946-50ul |
50 μg |
216.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHL-1/L1CAM-2 Polyclonal antibody specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for ELISASpecifications
| CHL-1/L1CAM-2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
| Produced in rabbits immunized with purified, recombinant Human CHL-1/L1CAM-2 (Accession#: AAI04919.1; Met 1-Gln 1080) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| 10752 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title