missing translation for 'onlineSavingsMsg'
Learn More

CHKB Polyclonal Antibody
GREENER_CHOICE

Product Code. 17160244 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17160244 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17160244 Supplier Thermo Scientific Supplier No. PA5112385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHKB Polyclonal Antibody

CHKB Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHKB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CHKB
Gene Accession No. O54783, O55229, Q9Y259
Gene Alias Chetk; CHKB; Chkl; choline kinase beta; Choline kinase-like protein; choline/ethanolamine kinase; choline/ethanolamine kinase beta; choline/ethanolamine kinase-b; choline/ethanolaminekinase; CK; Ck/Ek; Ck/Ek-beta; CKB; CKEKB; EK; EKB; ethanolamine kinase; Ethanolamine kinase beta; MDCMC
Gene Symbols CHKB
Host Species Rabbit
Immunogen Recombinant Human Choline/ethanolamine kinase protein (2-99AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1120, 12651, 29367
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.