missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: CEWVYDYTHE EWPFYKARPT DYPTQEQQLH FIRHYLAEAK KGETLSQEEQ RKLEEDLLVE VSRYALASHF FWGLWS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase, the initial enzyme in the sequence, plays a role in cell growth proliferation. A related protein, ChoKB (also, known as choline kinase beta), is a 395 amino acid enzyme that catalyzes the phosphorylation of choline by ATP in the presence of magnesium, thereby yielding phosphocholine and ADP. Like all choline kinases, ChoKB possesses ethanoalamine kinase activity and catalyzes the phosphorylation of ethanolamine. The gene encoding ChoKB is located less than 1 kb upstream of the CPT1B gene, suggesting that the ChoKB gene may regulate transcription CPT1B. In mice, mutations and/or deletions in the gene encoding ChoKB are the cause of hindlimb muscular dystrophy and neonatal bone deformity.
Specifications
Specifications
| Antigen | CHKB |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CHKB |
| Gene Accession No. | Q9Y259 |
| Gene Alias | Chetk; CHKB; Chkl; choline kinase beta; Choline kinase-like protein; choline/ethanolamine kinase; choline/ethanolamine kinase beta; choline/ethanolamine kinase-b; choline/ethanolaminekinase; CK; Ck/Ek; Ck/Ek-beta; CKB; CKEKB; EK; EKB; ethanolamine kinase; Ethanolamine kinase beta; MDCMC |
| Gene Symbols | CHKB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?