missing translation for 'onlineSavingsMsg'
Learn More

Chitosan oligosaccharide, MedChemExpress

Product Code. 16447287
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447287 5 g 5g
16411169 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447287 Supplier Medchem Express Supplier No. HY112108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. Chitosan oligosaccharide (COS) activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.

Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. Chitosan oligosaccharide (COS) activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.

Specifications

Chemical Name or Material Chitosan oligosaccharide
CAS 148411-57-8
Color Light Yellow
Purity Grade Notes Research
Quantity 5 g
Synonym COS
Solubility Information DMSO : 50 mg/mL (Need ultrasonic) ∣H2O : 50 mg/mL (Need ultrasonic)
SMILES [Chitosan oligosaccharide]
Percent Purity 91.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.