missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase-IN-2 hydrochloride, MedChemExpress
Description
Chitinase-IN-2 hydrochloride is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide
Specifications
Specifications
| Chemical Name or Material | Chitinase-IN-2 hydrochloride |
| Physical Form | Solid |
| CAS | 2070014-83-2 |
| Color | Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22ClN5O2S |
| Solubility Information | H2O : 50 mg/mL (115.76 mM; Need ultrasonic) ∣DMSO : ≥ 46 mg/mL (106.50 mM) |
| SMILES | O=C(C1=CC=CC2=C1C3=CC=C2N(C)C)N(CCNCC4=NN=C(C)S4)C3=O.Cl[H] |
| Molecular Weight (g/mol) | 431.94 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?