Learn More
Chitinase-IN-2 hydrochloride, MedChemExpress
Product Code. 16467476
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16467476
Brand: Medchem Express HY18599A
MedChemExpress Chitinase-IN-2 hydrochloride is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide; 50 uM/20uM compound concentration's inhibitory percentage are 98%/92% for chitinase/N- acetyl-hexosaminidase respectively.
Chitinase-IN-2 hydrochloride is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide
Specifications
Chemical Name or Material Chitinase-IN-2 hydrochloride
Physical Form Solid
CAS 2070014-83-2
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H22ClN5O2S
Solubility Information H2O : 50 mg/mL (115.76 mM; Need ultrasonic) ∣DMSO : ≥ 46 mg/mL (106.50 mM)
SMILES O=C(C1=CC=CC2=C1C3=CC=C2N(C)C)N(CCNCC4=NN=C(C)S4)C3=O.Cl[H]
Molecular Weight (g/mol) 431.94
Formula Weight 431.94
Percent Purity 99.16%
Grade Research