Learn More
Chicken anti-Rabbit IgG (H+L) Secondary Antibody, TRITC (Pre-adsorbed), Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75270
Product Code. 18252525
  • 166.00 EUR / 0.50mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
TRITC
Chicken
Affinity purified
RUO
Secondary
Rehydrate with 0.55 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store at 4C. Do not freeze.
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunomicroscopy
LYOPH
Flow Cytometry 1:20-1:2000, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000
Purified Rabbit IgG (H&L)
0.5 mg
Adaptive Immunity, Immunology
Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody has been pre-adsorbed against human and mouse IgG.
Rabbit
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions