Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, PAEK, 5μL
Product Code. 11543970
Change view
Click to view available options
Volume (Metric) Sample:
5 μL
Unit Size:
Each
Product Code. Volume (Metric) Sample unitSize
11543970 5 μL Each
1 options
Product Code. 11543970
Brand: Thermo Scientific™ CZSL5PK
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
  • 1/16″ fittings
  • 0.010″ ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Specifications
Volume (Metric) Sample 5 μL
Type Sample Injector Loop
Stator Material PEEK