Learn More
Brady™ Chemical-resistant Cryogenic Polypropylene Laboratory Labels for M6/M7 Printers
Product Code. 17878364
Change view
Click to view available options
Dimensions (LxW):
0.5 x 1 in.
0.6 x 1.3 in.
0.75 x 1 in.
1 x 1.5 in.
1.5 x 1.9 in.
2 x 0.25 in.
Height (Metric):
12.7 mm
15.24 mm
19.05 mm
25.4 mm
38.1 mm
50.8 mm
Unit Size:
Pack of 100
Pack of 250
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Height (Metric) unitSize
17878364 1 x 1.5 in. 25.4 mm Pack of 250
17861519 0.5 x 1 in. 12.7 mm Pack of 500
17868104 0.6 x 1.3 in. 15.24 mm Pack of 250
17838424 0.75 x 1 in. 19.05 mm Pack of 250
17858404 1.5 x 1.9 in. 38.1 mm Pack of 100
17858324 2 x 0.25 in. 50.8 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17878364 Supplier Brady™ Supplier No. 174101
Only null left
Add to basket
Add to basket
Polypropylene material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclave and other harsh environments.
  • Matte white polypropylene with acrylic adhesive is thermal transfer printable
  • Ideal for laboratory identification such as vials, centrifuge tubes and test tubes
  • Service temperature of -80°C to 70°C (-112°F to 158°F)
  • Made of B-7425 polypropylene material
Specifications
Autoclavable Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979, RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Polypropylene
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Equipment Labeling, General Identification, Laboratory Labeling, Temperature Indicating Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 250 Labels
Shape Rectangle
Temperature Range (English) -112°F to +158°F
Temperature Range (Metric) -80°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 1 x 1.5 in.
Length (English) 1 in.
Width (English) 1.5 in.
Width (Metric) 38.1 mm
Height (Metric) 25.4 mm
Alternate Part Number M6-31-7425