missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chebulagic acid, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
Chebulagic acid is a COX-LOX dual inhibitor isolated from the fruits of Terminalia chebula Retz, on angiogenesis. Chebulagic acid is a M2 serine to asparagine 31 mutation (S31N) inhibitor and influenza antiviral. Chebulagic acid also against SARS-CoV-2 viral replication with an EC50 of 9.76 μM.
Specifications
Specifications
| Chemical Name or Material | Chebulagic acid |
| CAS | 23094-71-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C41H30O27 |
| Solubility Information | DMSO : 100 mg/mL (104.75 mM; Need ultrasonic) |
| SMILES | OC(CC(C(O[C@H]([C@@H](COC(C1=CC(O)=C2O)=O)O[C@@H]3OC(C4=CC(O)=C(O)C(O)=C4)=O)[C@H](OC(C5=CC(O)=C(O)C(O)=C5C1=C2O)=O)[C@H]3OC6=O)=O)C(C7O)C(C6=CC(O)=C8O)=C8OC7=O)=O |
| Molecular Weight (g/mol) | 954.66 |
| Quantity | 2 mg |
| Formula Weight | 954.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction