missing translation for 'onlineSavingsMsg'
Learn More

Chebulagic acid, MedChemExpress

Product Code. 16480386
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16480386

Brand: Medchem Express HYN1996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chebulagic acid is a COX-LOX dual inhibitor isolated from the fruits of Terminalia chebula Retz, on angiogenesis. Chebulagic acid is a M2 serine to asparagine 31 mutation (S31N) inhibitor and influenza antiviral. Chebulagic acid also against SARS-CoV-2 viral replication with an EC50 of 9.76 μM.

Chebulagic acid is a COX-LOX dual inhibitor isolated from the fruits of Terminalia chebula Retz, on angiogenesis. Chebulagic acid is a M2 serine to asparagine 31 mutation (S31N) inhibitor and influenza antiviral. Chebulagic acid also against SARS-CoV-2 viral replication with an EC50 of 9.76 μM.

Specifications

Chemical Name or Material Chebulagic acid
CAS 23094-71-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C41H30O27
Solubility Information DMSO : 100 mg/mL (104.75 mM; Need ultrasonic)
SMILES OC(CC(C(O[C@H]([C@@H](COC(C1=CC(O)=C2O)=O)O[C@@H]3OC(C4=CC(O)=C(O)C(O)=C4)=O)[C@H](OC(C5=CC(O)=C(O)C(O)=C5C1=C2O)=O)[C@H]3OC6=O)=O)C(C7O)C(C6=CC(O)=C8O)=C8OC7=O)=O
Molecular Weight (g/mol) 954.66
Quantity 2 mg
Formula Weight 954.66
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.