missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total CHD1L.
In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
Specifications
Specifications
| Antigen | CHD1L |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | CHD1L |
| Gene Accession No. | Q86WJ1, Q9CXF7 |
| Gene Alias | 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792) |
| Gene Symbols | CHD1L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
