missing translation for 'onlineSavingsMsg'
Learn More

CGS-12066A Maleate Salt, TRC

Product Code. 30350952
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30350952 50 mg 50mg
30362149 25 mg -
2 options
This item is not returnable. View return policy

Product Code. 30350952

Brand: Toronto Research Chemicals TRCC29179350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1350965-83-1
Formula Weight 450.1515
InChI Formula InChI=1S/C17H17F3N4.C4H4O4/c1-22-7-9-23(10-8-22)16-15-3-2-6-24(15)14-5-4-12(17(18,19)20)11-13(14)21-16;5-3(6)1-2-4(7)8/h2-6,11H,7-10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
IUPAC Name (Z)-but-2-enedioic acid;4-(4-methylpiperazin-1-yl)-7-(trifluoromethyl)pyrrolo[1,2-a]quinoxaline
Molecular Formula C17H17F3N4 . C4H4O4
Molecular Weight (g/mol) 450.4
SMILES CN1CCN(CC1)c2nc3cc(ccc3n4cccc24)C(F)(F)F.OC(=O)\C=C/C(=O)O
Synonym CGS-12066A Maleate Salt, 4-(4-Methyl-1-piperazinyl)-7-(trifluoromethyl)-pyrrolo[1,2-a]quinoxaline (2Z)-2-Butenedioate
Chemical Name or Material 4-(4-Methyl-1-piperazinyl)-7-(trifluoromethyl)-pyrrolo[1,2-a]quinoxaline (2Z)-2-Butenedioate

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.