missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI 62 Antibody (OTI4C1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71896MFV610
This item is not returnable.
View return policy
Description
CGI 62 Monoclonal antibody specifically detects CGI 62 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| CGI 62 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 51101 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI4C1 | |
| Flow Cytometry | |
| C8orf70, CGI-62, chromosome 8 open reading frame 70, family with sequence similarity 164, member A, hypothetical protein LOC51101 | |
| Full length human recombinant protein of human FAM164A(NP_057094) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction