missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Cevimeline hydrochloride hemihydrate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
(-)-Cevimeline hydrochloride hemihydrate ((-)-SNI-2011), a novel muscarinic receptor agonist, is a candidate therapeutic drug for xerostomia in Sjogren's syndrome.
Specifications
Specifications
| Chemical Name or Material | (-)-Cevimeline hydrochloride hemihydrate |
| Physical Form | Solid |
| Color | Light Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H17NOS.HCl.1/2H2O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | (-)-SNI-2011(-)-AF102B hydrochloride hemihydrate |
| SMILES | C[C@@H](SC1)O[C@@]21CN3CCC2CC3.Cl.[0.5H2O] |
| Molecular Weight (g/mol) | 244.78 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction