missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine Dihydrochloride, TRC

Product Code. 17973955
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973955 100 mg 100mg
17992515 50 mg 50mg
17970715 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973955 Supplier Toronto Research Chemicals Supplier No. TRCC281100100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83881-52-1
Molecular Formula C21 H25 Cl N2 O3 . 2 Cl H
Molecular Weight (g/mol) 461.81
Synonym Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), Alercet, Alerid, Alerlisin, Cesta, Cetiriz, Cetirizine dihydrochloride, Cetirizine hydrochloride, Cetrak, Cetrine, Cetrizet, Cistamine, Epirizine, Formistin, Histazine, Histazine-1, Nosemin, P 071, Reactine, Riztec, Ryzen, Sancotec, Selitex, Tomazine, Triz, UCB-P 071, Virlix, Zeran, Zirtec, Zirtek, Zirtin, Zyrlex, Zyrtec, Zyrzine
IUPAC Name 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H
Formula Weight 460.1087
Percent Purity >95%
Chemical Name or Material Cetirizine Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.