missing translation for 'onlineSavingsMsg'
Learn More

Cesium chromate, 99.9% (metals basis)

Product Code. 11306868
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306868 100 g 100g
11396858 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306868 Supplier Thermo Scientific Alfa Aesar Supplier No. 013492.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13454-78-9 | CrCs2O4 | 381.80 g/mol

Cesium chromate is used to produce cesium vapor by reaction with silicon, boron, or titanium, which is used in the final stages of creating vacuum tubes. It is used in the production of photo-tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-78-9
Molecular Formula CrCs2O4
Molecular Weight (g/mol) 381.80
MDL Number MFCD00016050
InChI Key BROHICCPQMHYFY-UHFFFAOYSA-N
Synonym cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
PubChem CID 61613
IUPAC Name dicesium;dioxido(dioxo)chromium
SMILES [Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 100 g
UN Number UN3087
Solubility Information Soluble in water.
Formula Weight 381.81
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium chromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.