missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CES6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568885
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 100%.
Carboxylesterases that catalyzes the hydrolysis of pyrethroids pesticides. Hydrolyzes permethrin faster than cypermethrin.
Specifications
| CES6 | |
| Polyclonal | |
| Unconjugated | |
| Ces2a | |
| 9130231C15Rik; AI266984; carboxylesterase 2A; carboxylesterase 6; carboxylesterase isoenzyme; Ces2a; Ces6; Pyrethroid hydrolase Ces2a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 102022 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8QZR3 | |
| Ces2a | |
| synthetic peptide corresponding to a region of mouse CES6 (aa 251-300). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction