missing translation for 'onlineSavingsMsg'
Learn More

Cerivastatin Sodium Salt, TRC

Product Code. 30135341
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135341 5 mg 5mg
30133010 25 mg 25mg
30003341 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135341 Supplier Toronto Research Chemicals Supplier No. TRCC2770005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143201-11-0
Molecular Formula C26 H33 F N O5 . Na
Molecular Weight (g/mol) 481.53
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, sodium salt (1:1), (3R,5S,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, monosodium salt, (3R,5S,6E)- (9CI), 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]-3,5-dihydroxy-, monosodium salt, [S-[R*,S*-(E)]]-, BAY-w 6228, Baycol, Cerivastatin sodium, Lipobay, Rivastatin
IUPAC Name sodium;(E,3R,5S)-7-[4-(4-fluorophenyl)-5-(methoxymethyl)-2,6-di(propan-2-yl)pyridin-3-yl]-3,5-dihydroxyhept-6-enoate
SMILES [Na+].COCc1c(nc(C(C)C)c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c1c2ccc(F)cc2)C(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H34FNO5.Na/c1-15(2)25-21(11-10-19(29)12-20(30)13-23(31)32)24(17-6-8-18(27)9-7-17)22(14-33-5)26(28-25)16(3)4;/h6-11,15-16,19-20,29-30H,12-14H2,1-5H3,(H,31,32);/q;+1/p-1/b11-10+;/t19-,20-;/m1./s1
Formula Weight 481.224
Percent Purity >95%
Chemical Name or Material Cerivastatin, Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.